Läs mer
13-Ethylgon-5(10)en-3,17-dione, TRC
Produktkod. 30136617
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136617 50 mg
30006185 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136617 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE91859550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4222-96-2
Molekylformel C19 H26 O2
Molekylvikt (g/mol) 286.41
Synonym Gon-5(10)-ene-3,17-dione, 13-ethyl-, 13-Ethylgon-5(10)-ene-3,17-dione, 13-Ethylgon-5(10)en-3,17-dione, δ5(10)-Levodione
IUPAC-namn (8R,9S,13S,14S)-13-ethyl-1,2,4,6,7,8,9,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthrene-3,17-dione
LEDER CC[C@]12CC[C@H]3[C@@H](CCC4=C3CCC(=O)C4)[C@@H]1CCC2=O
Specifikationer
Kemiskt namn eller material delta5(10)-Levodione
Kvantitet 50 mg
InChI formel InChI=1S/C19H26O2/c1-2-19-10-9-15-14-6-4-13(20)11-12(14)3-5-16(15)17(19)7-8-18(19)21/h15-17H,2-11H2,1H3/t15-,16-,17+,19+/m1/s1
Formel vikt 286.1933