Läs mer
13-oxo-1,4,7,10-Tetraazabicyclo[8.2.1]tridecane-4,7-diacetic Acid, TRC
Produktkod. 17904045
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17904045 250 mg
17910995 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17904045 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT283945250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 229312-33-8
Molekylformel C13 H22 N4 O5
Molekylvikt (g/mol) 314.34
Synonym Gadoteridol Impurity 2
IUPAC-namn 2-[7-(carboxymethyl)-13-oxo-1,4,7,10-tetrazabicyclo[8.2.1]tridecan-4-yl]acetic acid
LEDER OC(=O)CN1CCN(CC(=O)O)CCN2CCN(CC1)C2=O
Specifikationer
Kemiskt namn eller material 13-oxo-1,4,7,10-Tetraazabicyclo[8.2.1]tridecane-4,7-diacetic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 250 mg
InChI formel InChI=1S/C13H22N4O5/c18-11(19)9-14-1-2-15(10-12(20)21)4-6-17-8-7-16(5-3-14)13(17)22/h1-10H2,(H,18,19)(H,20,21)
Formel vikt 314.159
Procent renhet >95%