Läs mer
13-Oxo-10-deacetyl Baccatin III, TRC
Produktkod. 30411600
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30411600 50 mg
30424857 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30411600 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO85011050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 92950-42-0
Molekylformel C29H34O10
Molekylvikt (g/mol) 542.57
Synonym (2aR,4S,4aS,6R,11S,12S,12aR,12bS)-12b-(Acetyloxy)-12-(benzoyloxy)-2a,4,4a,10,11,12,12a,12b-octahydro-4,6,11-trihydroxy-4a,8,13,13-tetramethyl-7,11-methano-1H-cyclodeca[3,4]benz[1,2-b]oxete-5,9(3H,6H)-dione, 10-Deacetyl-13-oxobaccatin III
LEDER CC1=C2[C@H](C(=O)[C@@]3([C@H](C[C@@H]4[C@]([C@H]3[C@@H]([C@@](C2(C)C)(CC1=O)O)OC(=O)C5=CC=CC=C5)(CO4)OC(=O)C)O)C)O
Specifikationer
Kemiskt namn eller material 13-Oxo-10-deacetyl Baccatin III
Kvantitet 50 mg
InChI formel InChI=1S/C29H34O10/c1-14-17(31)12-29(36)24(38-25(35)16-9-7-6-8-10-16)22-27(5,23(34)21(33)20(14)26(29,3)4)18(32)11-19-28(22,13-37-19)39-15(2)30/h6-10,18-19,21-22,24,32-33,36H,11-13H2,1-5H3/t18-,19+,21+,22-,24-,27+,28-,29+/m0/s1
Formel vikt 542.2152