Läs mer
13-OxoODE, TRC
Produktkod. 30425097
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30425097 5 mg
30408883 1 mg
30419612 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30425097 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO8584055MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 54739-30-9
Molekylformel C18H30O3
Molekylvikt (g/mol) 294.4
Synonym 13-KODE
IUPAC-namn 13-oxo-9Z,11E-octadecadienoic acid
LEDER CCCCCC(=O)/C=C\C=C\CCCCCCCC(=O)O
Specifikationer
Kemiskt namn eller material 13-OxoODE
Kvantitet 5 mg
InChI formel InChI=1S/C18H30O3/c1-2-3-11-14-17(19)15-12-9-7-5-4-6-8-10-13-16-18(20)21/h7,9,12,15H,2-6,8,10-11,13-14,16H2,1H3,(H,20,21)/b9-7-,15-12+