Läs mer
13-Oxyingenol Dodecanoate, TRC
Produktkod. 30425953
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30425953 10 mg
30413192 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30425953 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO87077010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 54706-70-6
Molekylformel C32H50O7
Molekylvikt (g/mol) 546.74
Synonym Dodecanoic Acid Ingenol Ester, [1aR-(1aα,2β,5β,5aβ,6β,8aα,9α,10aα)]-1,1a,2,5,5a,6,9,10-Octahydro-5,5a,6-trihydroxy-4-(hydroxymethyl)-1,1,7,9-tetramethyl-11-oxo-10aH-2,8a-methanocyclopenta[a]cyclopropa[e]cyclodecen-10a-yl Ester Dodecanoic Acid, 10aH-2,8a-Methanocyclopenta[a]cyclopropa[e]cyclodecene Dodecanoic Acid Deriv.
LEDER CCOc1ccc(cc1)C2(CC)C(=O)NC(=O)NC2=O
Specifikationer
Kemiskt namn eller material 13-Oxyingenol Dodecanoate
Kvantitet 10 mg
InChI formel InChI=1S/C32H50O7/c1-6-7-8-9-10-11-12-13-14-15-24(34)39-31-18-21(3)30-17-20(2)26(35)32(30,38)27(36)22(19-33)16-23(28(30)37)25(31)29(31,4)5/h16-17,21,23,25-27,33,35-36,38H,6-15,18-19H2,1-5H3/t21-,23+,25-,26+,27-,30+,31+,32-/m1/s1
Formel vikt 546.3557