Läs mer
(+)-13alpha-Hydroxylupanine, TRC
Produktkod. 30006765
Change view
Kvantitet:
1 mg
2.5 mg
10 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30006765 10 mg -
30005649 2.5 mg 2.5mg
30006542 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30006765 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH94379510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 15358-48-2
Molekylformel C15 H24 N2 O2
Molekylvikt (g/mol) 264.36
Synonym 7,14-Methano-2H,11H-dipyrido[1,2-a:1',2'-e][1,5]diazocin-11-one, dodecahydro-2-hydroxy-, (2S,7S,7aR,14S,14aS)-, 7,14-Methano-2H,11H-dipyrido[1,2-a:1',2'-e][1,5]diazocin-11-one, dodecahydro-2-hydroxy-, [2S-(2α,7β,7aβ,14β,14aα)]-, Lupanine, 13-hydroxy- (7CI), Lupanine, 13α-hydroxy- (8CI), Lupanine, hydroxy- (6CI), (2S,7S,7aR,14S,14aS)-Dodecahydro-2-hydroxy-7,14-methano-2H,11H-dipyrido[1,2-a:1',2'-e][1,5]diazocin-11-one, (+)-13-Hydroxylupanine, (+)-13α-Hydroxylupanine, 13-Hydroxylupanin, 13-Hydroxylupanine, 13α-Hydroxy-2-oxosparteine, 13α-Hydroxylupanine, 7,14-Methano-4H,6H-dipyrido[1,2-a:1',2'-e][1,5]diazocin-4-one, dodecahydro-9-hydroxy-, [7S-(7α,7aβ,9β,14α,14aα)]-, Hydroxylupanin, Hydroxylupanine, NSC 70827, Oxylupanine
LEDER O[C@H]1CCN2C[C@@H]3C[C@@H](CN4[C@@H]3CCCC4=O)[C@@H]2C1
Specifikationer
Kemiskt namn eller material (+)-13alpha-Hydroxylupanine
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C15H24N2O2/c18-12-4-5-16-8-10-6-11(14(16)7-12)9-17-13(10)2-1-3-15(17)19/h10-14,18H,1-9H2/t10-,11-,12-,13+,14-/m0/s1
Formel vikt 264.1838
Procent renhet >95%