Läs mer
14,15-Dehydro Budesonide, TRC
Produktkod. 30135779
Change view
Kvantitet:
10 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135779 50 mg
30135286 250 mg
17964475 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30135779 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD22930050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 131918-64-4
Molekylformel C25 H32 O6
Molekylvikt (g/mol) 428.518
Synonym 16α,17-[(1RS)-Butylidenebis(oxy)]-11β,21-dihydroxypregna-1,4,14-triene-3,20-dione, 14,15-Didehydrobudesonide, Budesonide Imp. E (EP), Pregna-1,4,14-triene-3,20-dione, 16,17-[butylidenebis(oxy)]-11,21-dihydroxy-, (11β,16α)-, (11β,16α)-16,17-[Butylidenebis(oxy)]-11,21-dihydroxypregna-1,4,14-triene-3,20-dione, 2H-Naphth[2',1':4,5]indeno[1,2-d][1,3]dioxole, pregna-1,4,14-triene-3,20-dione deriv., 16α,17-[Butylidenebis(oxy)]-11β,21-dihydroxypregna-1,4,14-triene-3,20-dione
IUPAC-namn (1R,4R,8S,9S,11S,12S,13R)-11-hydroxy-8-(2-hydroxyacetyl)-9,13-dimethyl-6-propyl-5,7-dioxapentacyclo[10.8.0.02,9.04,8.013,18]icosa-2,14,17-trien-16-one
LEDER CCCC1O[C@@H]2C=C3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@H]4[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO
Specifikationer
Kemiskt namn eller material 14,15-Dehydro Budesonide
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C25H32O6/c1-4-5-21-30-20-11-17-16-7-6-14-10-15(27)8-9-23(14,2)22(16)18(28)12-24(17,3)25(20,31-21)19(29)13-26/h8-11,16,18,20-22,26,28H,4-7,12-13H2,1-3H3/t16-,18-,20+,21?,22+,23-,24-,25+/m0/s1
Formel vikt 428.22
Procent renhet >95%