Läs mer
14,15-Dehydro Triamcinolone Acetonide, TRC
Produktkod. 18083150
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18083150 25 mg
18043050 10 mg
18000860 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 18083150 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD23057025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1260149-96-9
Molekylformel C24 H29 F O6
Molekylvikt (g/mol) 432.48
Synonym 9-Fluoro-11β,21-dihydroxy-16α,17-(1-methylethylidenedioxy)pregna-1,4,14-triene-3,20-dione (δ14-Triamcinolone Acetonide)
IUPAC-namn (1S,4R,8S,9S,11S,12R,13S)-12-fluoro-11-hydroxy-8-(2-hydroxyacetyl)-6,6,9,13-tetramethyl-5,7-dioxapentacyclo[10.8.0.02,9.04,8.013,18]icosa-2,14,17-trien-16-one
LEDER CC1(C)O[C@@H]2C=C3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@@]4(F)[C@@H](O)C[C@]3(C)[C@@]2(O1)C(=O)CO
Specifikationer
Kemiskt namn eller material 14,15-Dehydro Triamcinolone Acetonide
Kvantitet 25 mg
InChI formel InChI=1S/C24H29FO6/c1-20(2)30-19-10-16-15-6-5-13-9-14(27)7-8-21(13,3)23(15,25)17(28)11-22(16,4)24(19,31-20)18(29)12-26/h7-10,15,17,19,26,28H,5-6,11-12H2,1-4H3/t15-,17-,19+,21-,22-,23-,24+/m0/s1
Formel vikt 432.1948