Läs mer
15,16-Deshydroxy 3-O-Benzyl Estetrol 17-Acetate, TRC
Produktkod. 30358069
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30358069 25 mg
1 options
Produktkod. 30358069 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD29002525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 15,16-Deshydroxy 3-O-Benzyl Estetrol 17-Acetate
CAS 690996-25-9
Molekylformel C27H30O3
Kvantitet 25 mg
Synonym (17beta)-3-(Phenylmethoxy)-estra-1,3,5(10),15-tetraen-17-ol Acetate
InChI formel InChI=1S/C27H30O3/c1-18(28)30-26-13-12-25-24-10-8-20-16-21(29-17-19-6-4-3-5-7-19)9-11-22(20)23(24)14-15-27(25,26)2/h3-7,9,11-13,16,23-26H,8,10,14-15,17H2,1-2H3/t23-,24-,25+,26-,27+/m1/s1
LEDER CC(=O)O[C@@H]1C=C[C@H]2[C@@H]3CCc4cc(OCc5ccccc5)ccc4[C@H]3CC[C@]12C
Molekylvikt (g/mol) 402.53