missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, for HPLC-MS
3550.67 SEK valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
1 L
2.5 L
Description
Acetonitrile, for HPLC-MS
Specifications
Packaging | Amber glass bottle |
Quantity | 2.5 L |
Color | Colorless |
Molecular Weight (g/mol) | 41.05 |
Formula Weight | 41.05 g/mol |
Vapor Pressure | 97 mbar at 20°C |
Viscosity | 0.36 cp at 20°C |
Vapor Density | 1.42 |
Melting Point | -46°C |
Boiling Point | 81°C |
Show More |
Safety and Handling
Product Identifier
- Acetonitrile
- Danger
- Flammable liquid Category 2
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'