Learn More
SI Analytics™ ProcessLine Combination Electrode
SI Analytics™ ProcessLine Combination Electrode is a low maintenance pH electrode with a temperature sensor.
Brand:  SI Analytics™ 285113550
Additional Details : Weight : 1.50000kg
 View more versions of this product
Product Code. 15395866
  • 4380.00 SEK / Each
Estimated Shipment: 06-05-2024
to see stock.
Add to basket
Description
Description
  • Low-maintenance
  • pH combination electrode with Pt 1000 or PT 100
  • Glass shaft
  • Silamid™ reference
  • 2 hole diaphragm
  • DuraLid reference system
  • Sphere membrane
  • H glassVP screw plug head
Specifications
Specifications
Screw Plug Head
0 to 14
0°C to 130°C
12 mm
H-Glass
pH Combination Electrode
120 mm