Learn More
Perchloric acid solution, Fluka™
Volumetric, 0.1 M HClO4 in glacial acetic acid (0.1N), for titration in non-aqueous medium
Brand: Honeywell Fluka 35418-500ML
Additional Details : CAS Number : 7601-90-3 Weight : 0.52500kg Transport : UN number : 2920 Chem class : 8 Pack group : II
Description
Honeywell's high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
Specifications
Acidic (as aqueous solution) | |
Colorless | |
119°C (1.013 hPa) (The information regarding the boiling point is that of the solvent.) | |
500 mL |
Perchloric acid solution | |
7601-90-3,64-19-7 | |
Glass Bottle | |
HClO4 |
Safety and Handling
- Perchloric acid solution
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H226-Flammable liquid and vapour.
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P234-Keep only in original container.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : Acetic acid, perchloric acid