missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Phenolphthalein solution, Indicator solution, Honeywell Fluka™
Handla allt Honeywell Fluka ProdukterBeskrivning
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
| Kemiskt namn eller material | Phenolphthalein solution |
| Namnnotering | Indicator solution |
| Kokpunkt | 78°C to 79°C |
| CAS | 77-09-8, 64-17-5 |
| CAS Max % | 0.1000%, 99.9000% |
| Molekylformel | C20H14O4 |
| Linjär formel | C20H14O4 |
| MDL-nummer | MFCD00005913 |
| UN-nummer | UN1170 |
| Beilstein | 284423 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?