Läs mer
16,17-Dehydro Capsaicin, TRC
Produktkod. 30350277
Change view
Kvantitet:
0.5 mg
1 mg
5 mg
Förpackningsstorlek:
0.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30350277 0.5 mg 0.5mg
30364292 5 mg -
30366448 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30350277 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD2293250.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 509101-57-9
Molekylformel C18H25NO3
Molekylvikt (g/mol) 303.4
Synonym (6E)-N-[(4-Hydroxy-3-methoxyphenyl)methyl]-8-methyl-6,8-nonadienamide
IUPAC-namn (6E)-N-[(4-hydroxy-3-methoxyphenyl)methyl]-8-methylnona-6,8-dienamide
LEDER COc1cc(CNC(=O)CCCC\C=C\C(=C)C)ccc1O
Specifikationer
Kemiskt namn eller material 16,17-Dehydro Capsaicin
Kvantitet 0.5 mg
InChI formel InChI=1S/C18H25NO3/c1-14(2)8-6-4-5-7-9-18(21)19-13-15-10-11-16(20)17(12-15)22-3/h6,8,10-12,20H,1,4-5,7,9,13H2,2-3H3,(H,19,21)/b8-6+
Formel vikt 303.1834