Läs mer
16-b-Homo Betamethasone Phosphate, TRC
Produktkod. 30407122
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30407122 10 mg
1 options
Produktkod. 30407122 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH15462010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 16-b-Homo Betamethasone Phosphate
CAS 1202002-02-5
Molekylformel C22H30FO8P
Kvantitet 10 mg
Synonym (2S,3S,4aS,4bS,10aS,10bR,11S,12aS)-10b-Fluoro-3,4,4a,5,6,10a,10b,11,12,12a-decahydro-2,11-dihydroxy-3,10a,12a-trimethyl-2-[(phosphonooxy)methyl]-1,8(2H,4bH)-chrysenedione, ((2S,3S,4aS,4bS,10aS,10bR,11S,12aS)-10b-fluoro-2,11-dihydroxy-3,10a,12a-trimethyl-1,8-dioxo-1,2,3,4,4a,4b,5,6,8,10a,10b,11,12,12a-tetradecahydrochrysen-2-yl)methyl dihydrogen phosphate, betamethasone impurity 86
InChI formel InChI=1S/C22H30FO8P/c1-12-8-16-15-5-4-13-9-14(24)6-7-20(13,3)22(15,23)17(25)10-19(16,2)18(26)21(12,27)11-31-32(28,29)30/h6-7,9,12,15-17,25,27H,4-5,8,10-11H2,1-3H3,(H2,28,29,30)/t12-,15-,16-,17-,19-,20-,21+,22-/m0/s1
LEDER F[C@@]12[C@]([C@]3([C@](C)(C[C@@H]1O)C(=O)[C@](COP(=O)(O)O)(O)[C@@H](C)C3)[H])(CCC=4[C@]2(C)C=CC(=O)C4)[H]
IUPAC-namn ((2S,3S,4aS,4bS,10aS,10bR,11S,12aS)-10b-fluoro-2,11-dihydroxy-3,10a,12a-trimethyl-1,8-dioxo-1,2,3,4,4a,4b,5,6,8,10a,10b,11,12,12a-tetradecahydrochrysen-2-yl)methyl dihydrogen phosphate
Molekylvikt (g/mol) 472.44
Formel vikt 472.17