Läs mer
16-Deacetylfusidic Acid gamma-Lactone, TRC
Produktkod. 30359004
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30359004 100 mg
1 options
Produktkod. 30359004 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD199075100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Deacetylfusidic Acid 21,16-Lactone
CAS 4701-54-6
Molekylformel C29 H44 O4
Kvantitet 100 mg
Synonym 29-Nordammara-17(20),24-dien-21-oic acid, 3,11,16-trihydroxy-, γ-lactone, (3α,4α,8α,9β,11α,13α,14β,16β)-, 29-Nor-8α,9β,13α,14β-dammara-17(20),24-dien-21-oic acid, 3α,11α,16β-trihydroxy-, γ-lactone (7CI,8CI), 16-Deacetylfusidic acid γ-lactone, 16-O-Deacetylfusidic acid lactone, ent-(17Z)-3β,11β-Dihydroxy-4β,8,14-trimethyl-18-nor-5β,10α-cholesta-17(20),24-dieno-21(16α)-lactone, Deacetylfusidic acid 21,16-lactone
InChI formel InChI=1S/C29H44O4/c1-16(2)8-7-9-18-24-20-14-22(31)25-27(4)12-11-21(30)17(3)19(27)10-13-28(25,5)29(20,6)15-23(24)33-26(18)32/h8,17,19-23,25,30-31H,7,9-15H2,1-6H3/t17-,19-,20-,21+,22+,23-,25-,27-,28-,29-/m0/s1
LEDER C[C@@H]1[C@H](O)CC[C@@]2(C)[C@H]1CC[C@@]3(C)[C@H]2[C@H](O)C[C@H]4C5=C(CCC=C(C)C)C(=O)O[C@H]5C[C@]34C
IUPAC-namn (1S,2S,4S,9R,11R,12S,13S,16R,17S,18S)-11,16-dihydroxy-1,2,13,17-tetramethyl-7-(4-methylpent-3-enyl)-5-oxapentacyclo[10.8.0.02,9.04,8.013,18]icos-7-en-6-one
Molekylvikt (g/mol) 456.6573
Formel vikt 456.324