Läs mer
16-Dehydro Pregnenolone Acetate, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | 16-Dehydropregnenoloneacetate |
| CAS | 979-02-2 |
| Molekylformel | C23 H32 O3 |
| Kvantitet | 10 g |
| Synonym | (3β)-3-(Acetyloxy)-pregna-5,16-dien-20-one, Pregna-5,16-dien-20-one, 3β-hydroxy-, acetate (6CI,8CI), (-)-16-Dehydropregnenolone acetate, 16,17-Didehydropregnenolone acetate, 16-DPA, 16-Dehydropregnenolone acetate, 16-Dehydropregnenolone-3β-acetate, 20-Oxopregna-5,16-dien-3β-yl acetate, 3β-Acetoxypregna-5,16-dien-20-one, 3β-Acetoxypregna-5,16-diene-20-one, 3β-Acetyloxy-pregna-5,16-dien-20-one, Dehydropregnenolone acetate, NSC 37741, Pregnadienolone acetate, (-)-16-Dehydropregnenolone acetate, 16,17-Didehydropregnenolone acetate, 16-DPA, 16-Dehydropregnenolone acetate, Dehydropregnenolone acetate, NSC 37741, Pregnadienolone acetate, (3beta)-3-(Acetyloxy)-pregna-5,16-dien-20-one, Pregna-5,16-dien-20-one, 3beta-hydroxy-, acetate (6CI,8CI), 16-Dehydropregnenolone-3beta-acetate, 20-Oxopregna-5,16-dien-3beta-yl acetate, 3beta-Acetoxypregna-5,16-dien-20-one, 3beta-Acetoxypregna-5,16-diene-20-one, 3beta-Acetyloxy-pregna-5,16-dien-20-one |
| InChI formel | InChI=1S/C23H32O3/c1-14(24)19-7-8-20-18-6-5-16-13-17(26-15(2)25)9-11-22(16,3)21(18)10-12-23(19,20)4/h5,7,17-18,20-21H,6,8-13H2,1-4H3/t17-,18-,20-,21-,22-,23+/m0/s1 |
| LEDER | CC(=O)O[C@H]1CC[C@]2(C)[C@H]3CC[C@@]4(C)[C@@H](CC=C4C(=O)C)[C@@H]3CC=C2C1 |
| IUPAC-namn | [(3S,8R,9S,10R,13S,14S)-17-acetyl-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15-decahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate |
| Molekylvikt (g/mol) | 356.5 |
| Formel vikt | 356.2351 |