Läs mer
16-Epi-deacetyl-fusidic Acid, TRC
Produktkod. 30009517
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009517 25 mg
30011572 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30009517 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE58300025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5951-83-7
Molekylformel C29 H46 O5
Molekylvikt (g/mol) 474.6725
Synonym 29-Nordammara-17(20),24-dien-21-oic acid, 3,11,16-trihydroxy-, (3α,4α,8α,9β,11α,13α,14β,16α,17Z)-, 16-Epifusidic acid, deacetyl- (7CI), 29-Nor-8α,9β,13α,14β-dammara-17(20),24-dien-21-oic acid, 3α,11α,16α-trihydroxy-, (Z)- (8CI), (3α,4α,8α,9β,11α,13α,14β,16α,17Z)-3,11,16-Trihydroxy-29-nordammara-17(20),24-dien-21-oic acid, 16-Epideacetylfusidic acid, NSC 319631, ent-(17Z)-3β,11β,16β-Trihydroxy-4β,8,14-trimethyl-18-nor-5β,10α-cholesta-17(20),24-dien-21-oic acid, 16-epi-Deacetylfusidic acid
IUPAC-namn (2Z)-6-methyl-2-[(3R,4S,5S,8S,9S,10S,11R,13R,14S,16R)-3,11,16-trihydroxy-4,8,10,14-tetramethyl-2,3,4,5,6,7,9,11,12,13,15,16-dodecahydro-1H-cyclopenta[a]phenanthren-17-ylidene]hept-5-enoic acid
LEDER C[C@@H]1[C@H](O)CC[C@@]2(C)[C@H]1CC[C@@]3(C)[C@H]2[C@H](O)C[C@H]4\C(=C(/CCC=C(C)C)\C(=O)O)\[C@H](O)C[C@]34C
Specifikationer
Kemiskt namn eller material 16-epi-Deacetylfusidic Acid
Kvantitet 25 mg
InChI formel InChI=1S/C29H46O5/c1-16(2)8-7-9-18(26(33)34)24-20-14-22(31)25-27(4)12-11-21(30)17(3)19(27)10-13-28(25,5)29(20,6)15-23(24)32/h8,17,19-23,25,30-32H,7,9-15H2,1-6H3,(H,33,34)/b24-18-/t17-,19-,20-,21+,22+,23+,25-,27-,28-,29-/m0/s1
Formel vikt 474.3345