missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
16α-Homo Dexamethasone Phosphate, TRC
Specifications
Specifications
| Kemiskt namn eller material | 16Alpha-Homo Dexamethasone Phosphate |
| Analyt- eller komponentnamn | 16Alpha-Homo Dexamethasone Phosphate |
| Frakt skick | Room Temperature |
| Molekylformel | C22H30FO8P |
| Synonym | ((4aS,4bS,10aS,10bR,11S,12aS)-10b-Fluoro-1,11-dihydroxy-3,10a,12a-trimethyl-2,8-dioxo-1,2,3,4,4a,4b,5,6,8,10a,10b,11,12,12a-tetradecahydrochrysen-1-yl)methyl Dihydrogen Phosphate, Dexamethasone Phosphate Impurity |
| InChI formel | InChI=1S/C22H30FO8P/c1-12-8-16-15-5-4-13-9-14(24)6-7-19(13,2)22(15,23)17(25)10-20(16,3)21(27,18(12)26)11-31-32(28,29)30/h6-7,9,12,15-17,25,27H,4-5,8,10-11H2,1-3H3,(H2,28,29,30)/t12?,15-,16-,17-,19-,20-,21?,22-/m0/s1 |
| Lösningstyp | Neat |
| LEDER | C[C@]12C(CC[C@@]3([C@@]1([C@]([H])(C[C@]4([C@]3(CC(C(C4(COP(O)(O)=O)O)=O)C)[H])C)O)F)[H])=CC(C=C2)=O |
| IUPAC-namn | ((4aS,4bS,10aS,10bR,11S,12aS)-10b-fluoro-1,11-dihydroxy-3,10a,12a-trimethyl-2,8-dioxo-1,2,3,4,4a,4b,5,6,8,10a,10b,11,12,12a-tetradecahydrochrysen-1-yl)methyl dihydrogen phosphate |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?