missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Ribonuclease UK114 Protein

A DNA sequence encoding the Homo sapiens (Human) Ribonuclease UK114, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6182-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16011390

  • 22640.00 SEK / 1mg
Estimated Shipment: 13-11-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_005827.1
Ribonuclease UK114 Protein
Research Use Only
HRSP12
Recombinant Protein
E. coli
SSLIRRVIST AKAPGAIGPY SQAVLVDRTI YISGQIGMDP SSGQLVSGGV AEEAKQALKN MGEILKAAGC DFTNVVKTTV LLADINDFNT VNEIYKQYFK SNFPARAAYQ VAALPKGSRI EIEAVAIQGP LTTASL
Greater than 90% as determined by SDS-PAGE.
10247
1 mg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers