Läs mer
IVL, Mouse, Clone: SY8, Abnova™
Mouse monoclonal antibody raised against IVL.
Brand: Abnova MAB7025.100ug
Beskrivning
Involucrin, a component of the keratinocyte crosslinked envelope, is found in the cytoplasm and crosslinked to membrane proteins by transglutaminase. This gene is mapped to 1q21, among calpactin I light chain, trichohyalin, profillaggrin, loricrin, and calcyclin. [provided by RefSeq
Specifikationer
| IVL | |
| Monoclonal | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| IVL | |
| Mouse | |
| 100 μg | |
| Primary | |
| Keratinocytes of epidermis and other stratified squamous epithelia. Predicted molecular weight 68 KDa. Reacts weakly with dog involucrin. | |
| Store at 4°C for short term. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG1 |
| Immunohistochemistry, Immunoprecipitation, Western Blot | |
| SY8 | |
| Mouse monoclonal antibody raised against IVL. | |
| In PBS | |
| IVL | |
| Human IVL. | |
| RUO | |
| 3713 | |
| Canine, Human, Pig, Primate | |
| Liquid |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.