Läs mer
(16Alpha)-21-(Acetyloxy)-16-methyl-pregna-1,4,9(11)-triene-3,20-dione, TRC
Produktkod. 30313962
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30313962 5 mg
30313797 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30313962 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA1874255MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (16alpha)-21-(Acetyloxy)-16-methyl-pregna-1,4,9(11)-triene-3,20-dione
CAS 4258-83-7
Molekylformel C24H30O4
Kvantitet 5 mg
Synonym 21-Acetoxy-16alpha-methylpregna-1,4,9-(11)-triene-3,20-dione
InChI formel InChI=1S/C24H30O4/c1-14-11-20-18-6-5-16-12-17(26)7-9-23(16,3)19(18)8-10-24(20,4)22(14)21(27)13-28-15(2)25/h7-9,12,14,18,20,22H,5-6,10-11,13H2,1-4H3/t14-,18-,20+,22?,23+,24+/m1/s1
LEDER C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)C3=CC[C@]2(C)C1C(=O)COC(=O)C
IUPAC-namn [2-oxo-2-[(8S,10S,13S,14S,16R)-10,13,16-trimethyl-3-oxo-6,7,8,12,14,15,16,17-octahydrocyclopenta[a]phenanthren-17-yl]ethyl] acetate
Molekylvikt (g/mol) 382.49
Formel vikt 382.2144