Läs mer
16alpha-Hydroxy Estrone, TRC
Produktkod. 18041530
Change view
Kvantitet:
1 mg
2.5 mg
5 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
18041530 1 mg -
18090810 2.5 mg 2.5mg
18081520 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 18041530 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH9419001MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 566-76-7
Molekylformel C18 H22 O3
Molekylvikt (g/mol) 286.37
Synonym (16α)-3,16-Dihydroxyestra-1,3,5(10)-trien-17-one, Estra-1,3,5(10)-trien-17-one, 3,16α-dihydroxy- (8CI), (16α)-3,16-Dihydroxyestra-1,3,5(10)-trien-17-one, 16α-Hydroxyestrone, 3,16α-Dihydroxy-1,3,5(10)-estratrien-17-one, Estra-1,3,5(10)-triene-3,16α-diol-17-one, Estriol Imp. H (EP)
IUPAC-namn (8R,9S,13S,14S,16R)-3,16-dihydroxy-13-methyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-17-one
LEDER C[C@]12CC[C@H]3[C@@H](CCc4cc(O)ccc34)[C@@H]1C[C@@H](O)C2=O
Specifikationer
Kemiskt namn eller material 16a-Hydroxy Estrone
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C18H22O3/c1-18-7-6-13-12-5-3-11(19)8-10(12)2-4-14(13)15(18)9-16(20)17(18)21/h3,5,8,13-16,19-20H,2,4,6-7,9H2,1H3/t13-,14-,15+,16-,18+/m1/s1
Formel vikt 286.1569
Procent renhet >95%