Läs mer
16Alpha-Methyl-11-oxo Prednisolone, TRC
Produktkod. 18014040
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18014040 10 mg
18093120 25 mg
18062930 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 18014040 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM29535010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2036-77-3
Molekylformel C22 H28 O5
Molekylvikt (g/mol) 372.45
Synonym Pregna-1,4-diene-3,11,20-trione, 17,21-dihydroxy-16-methyl-, (16α)-, Pregna-1,4-diene-3,11,20-trione, 17,21-dihydroxy-16α-methyl- (6CI,7CI,8CI), (16α)-17,21-Dihydroxy-16-methylpregna-1,4-diene-3,11,20-trione, 17,21-Dihydroxy-16α-methylpregna-1,4-diene-3,11,20-trione, Dexamethasone Imp. J (EP)
IUPAC-namn (8S,9S,10R,13S,14S,16R,17R)-17-hydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-6,7,8,9,12,14,15,16-octahydrocyclopenta[a]phenanthrene-3,11-dione
LEDER C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3C(=O)C[C@]2(C)[C@@]1(O)C(=O)CO
Specifikationer
Kemiskt namn eller material 17,21-Dihydroxy-16a-methylpregna-1,4-diene-3,11,20-trione
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C22H28O5/c1-12-8-16-15-5-4-13-9-14(24)6-7-20(13,2)19(15)17(25)10-21(16,3)22(12,27)18(26)11-23/h6-7,9,12,15-16,19,23,27H,4-5,8,10-11H2,1-3H3/t12-,15+,16+,19-,20+,21+,22+/m1/s1
Formel vikt 372.1937
Procent renhet >95%