Läs mer
16Alpha-Methyl-9,11-dehydro Prednisolone, TRC
Produktkod. 30009570
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009570 10 mg
30005040 25 mg
30012688 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30009570 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM29530010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 13209-41-1
Molekylformel C22 H28 O4
Molekylvikt (g/mol) 356.46
Synonym 17,21-Dihydroxy-16α-methyl-pregna-1,4,9(11)-triene-3,20-dione, Pregna-1,4,9(11)-triene-3,20-dione, 17,21-dihydroxy-16α-methyl- (6CI,7CI,8CI), 17α,21-Dihydroxy-16α-methylpregna-1,4,9(11)-triene-3,20-dione, VBP 15, Vamorolone
IUPAC-namn (8S,10S,13S,14S,16R,17R)-17-hydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-7,8,12,14,15,16-hexahydro-6H-cyclopenta[a]phenanthren-3-one
LEDER C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)C3=CC[C@]2(C)[C@@]1(O)C(=O)CO
Specifikationer
Kemiskt namn eller material 17,21-Dihydroxy-16alpha-methylpregna-1,4,9(11)-triene-3,20-dione
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C22H28O4/c1-13-10-18-16-5-4-14-11-15(24)6-8-20(14,2)17(16)7-9-21(18,3)22(13,26)19(25)12-23/h6-8,11,13,16,18,23,26H,4-5,9-10,12H2,1-3H3/t13-,16-,18+,20+,21+,22+/m1/s1
Formel vikt 356.1988
Procent renhet >95%