Läs mer
(16b)-21-(Acetyloxy)-2'-methyl-5'H-Pregna-1,4-dieno[17,16-d]oxazole-3,11,20-trione, TRC
Produktkod. 30323599
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30323599 1 g
1 options
Produktkod. 30323599 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA1737851G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (16beta)-21-(Acetyloxy)-2'-methyl-5'H-Pregna-1,4-dieno[17,16-d]oxazole-3,11,20-trione
CAS 13649-84-8
Molekylformel C25H29NO6
Kvantitet 1 g
Synonym 21-Hydroxy-2'-methyl-5'betaH-Pregna-1,4-dieno[17,16-d]oxazole-3,11,20-trione Acetate (Ester), Deflazacort Impurity
InChI formel InChI=1S/C25H29NO6/c1-13-26-25(20(30)12-31-14(2)27)21(32-13)10-18-17-6-5-15-9-16(28)7-8-23(15,3)22(17)19(29)11-24(18,25)4/h7-9,17-18,21-22H,5-6,10-12H2,1-4H3/t17-,18-,21+,22+,23-,24-,25+/m0/s1
LEDER CC(=O)OCC(=O)[C@@]12N=C(C)O[C@@H]1C[C@H]3[C@@H]4CCC5=CC(=O)C=C[C@]5(C)[C@H]4C(=O)C[C@]23C
Molekylvikt (g/mol) 439.5
Formel vikt 439.1995