Läs mer
17-Amino Geldanamycin, TRC
Produktkod. 30311353
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30311353 10 mg
30314583 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30311353 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA60970010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 17-Amino Geldanamycin
CAS 64202-81-9
Molekylformel C28 H39 N3 O8
Kvantitet 10 mg
Synonym 17-Aminogeldanamycin
InChI formel InChI=1S/C28H39N3O8/c1-14-10-18-23(29)20(32)13-19(25(18)34)31-27(35)15(2)8-7-9-21(37-5)26(39-28(30)36)17(4)12-16(3)24(33)22(11-14)38-6/h7-9,12-14,16,21-22,24,26,33H,10-11,29H2,1-6H3,(H2,30,36)(H,31,35)/b9-7-,15-8+,17-12+/t14-,16+,21+,22+,24-,26+/m1/s1
LEDER CO[C@H]1C[C@H](C)CC2=C(N)C(=O)C=C(NC(=O)\C(=C\C=C/[C@H](OC)[C@@H](OC(=O)N)\C(=C\[C@H](C)[C@H]1O)\C)\C)C2=O
IUPAC-namn [(3R,5S,6R,7S,8E,10S,11S,12Z,14E)-21-amino-6-hydroxy-5,11-dimethoxy-3,7,9,15-tetramethyl-16,20,22-trioxo-17-azabicyclo[16.3.1]docosa-1(21),8,12,14,18-pentaen-10-yl] carbamate
Molekylvikt (g/mol) 545.62
Formel vikt 545.2737