missing translation for 'onlineSavingsMsg'
Läs mer

17-Dehydroxy Prednisolone, TRC

Produktkod. 30354881
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30354881 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30354881 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD230025100MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material 17-Dehydroxy Prednisolone
CAS 13479-38-4
Molekylformel C21 H28 O4
Kvantitet 100 mg
Synonym Pregna-1,4-diene-3,20-dione, 11,21-dihydroxy-, (11β)-, Pregna-1,4-diene-3,20-dione, 11β,21-dihydroxy- (7CI,8CI), (11β)-11,21-Dihydroxypregna-1,4-diene-3,20-dione, 1,2-Didehydrocorticosterone, 1-Dehydrocorticosterone, 11β,21-Dihydroxypregna-1,4-diene-3,20-dione, 17-Deoxyprednisolone, δ1-Corticosterone
InChI formel InChI=1S/C21H28O4/c1-20-8-7-13(23)9-12(20)3-4-14-15-5-6-16(18(25)11-22)21(15,2)10-17(24)19(14)20/h7-9,14-17,19,22,24H,3-6,10-11H2,1-2H3/t14-,15-,16+,17-,19+,20-,21-/m0/s1
LEDER C[C@]12C[C@H](O)[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@@H]2C(=O)CO
IUPAC-namn (8S,9S,10R,11S,13S,14S,17S)-11-hydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-3-one
Molekylvikt (g/mol) 344.44
Formel vikt 344.1988
Produkttitel
Välj ett problem

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.