Läs mer
17-Deoxy Cortienic Acid, TRC
Produktkod. 30008592
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008592 25 mg
30008053 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30008592 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD23269525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2394-25-4
Molekylformel C20 H28 O4
Molekylvikt (g/mol) 332.43
Synonym (8S,9S,10R,11S,13S,14S,17S)-11-hydroxy-10,13-dimethyl-3-oxo-2,3,6,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthrene-17-carboxylic acid
IUPAC-namn (8S,9S,10R,11S,13S,14S,17S)-11-hydroxy-10,13-dimethyl-3-oxo-1,2,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthrene-17-carboxylic acid
LEDER C[C@@]12[C@@]3([C@]([C@]4([C@](C)(C[C@@H]3O)[C@@H](C(O)=O)CC4)[H])(CCC1=CC(=O)CC2)[H])[H]
Specifikationer
Kemiskt namn eller material Corticosterone Impurity 2
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C20H28O4/c1-19-8-7-12(21)9-11(19)3-4-13-14-5-6-15(18(23)24)20(14,2)10-16(22)17(13)19/h9,13-17,22H,3-8,10H2,1-2H3,(H,23,24)/t13-,14-,15+,16-,17+,19-,20-/m0/s1
Formel vikt 332.1988
Procent renhet >95%