Läs mer
17-epi-Estradiol Valerate (17alpha-Estradiol Valerate), Mikromol™
Produktkod. 30049375
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30049375 25 mg
30050861 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30049375 Leverantör Mikromol Leverantörsnummer MM0209.030025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
17-epi-Estradiol Valerate (17alpha-Estradiol Valerate)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 17-epi-Estradiol Valerate
CAS 182624-54-0
Analyt- eller komponentnamn 17-epi-Estradiol Valerate
Frakt skick Room Temperature
Molekylformel C23 H32 O3
Certifikat och efterlevnad ISO 9001
Synonym 17-epi-Estradiol Valerate, 17alpha-Estradiol Valerate
InChI formel InChI=1S/C23H32O3/c1-3-4-5-22(25)26-21-11-10-20-19-8-6-15-14-16(24)7-9-17(15)18(19)12-13-23(20,21)2/h7,9,14,18-21,24H,3-6,8,10-13H2,1-2H3/t18-,19-,20+,21-,23+/m1/s1
LEDER CCCCC(=O)O[C@@H]1CC[C@H]2[C@@H]3CCc4cc(O)ccc4[C@H]3CC[C@]12C
IUPAC-namn [(8R,9S,13S,14S,17R)-3-hydroxy-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-17-yl] pentanoate
Molekylvikt (g/mol) 356.5
Kvantitet 25 mg
Formel vikt 356.2351 g/mol