Läs mer
17-epi-Ethynyl Estradiol, TRC
Produktkod. 18062240
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18062240 10 mg
18033240 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18062240 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE68509510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4717-38-8
Molekylformel C20 H24 O2
Molekylvikt (g/mol) 296.4
Synonym 19-Norpregna-1,3,5(10)-trien-20-yne-3,17-diol, 17-epi-Ethynylestradiol, 17α-Estradiol, 17-ethynyl-, 17β-Ethynylestradiol, 19-Norpregna-1,3,5(10)-trien-20-yne-3,17-diol (17β-Ethinylestradiol), Ethinylestradiol Imp. A (EP)
IUPAC-namn (8R,9S,13S,14S,17S)-17-ethynyl-13-methyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthrene-3,17-diol
LEDER C[C@]12CC[C@H]3[C@@H](CCc4cc(O)ccc34)[C@@H]1CC[C@]2(O)C#C
Specifikationer
Kemiskt namn eller material 17beta-Ethinylestradiol
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C20H24O2/c1-3-20(22)11-9-18-17-6-4-13-12-14(21)5-7-15(13)16(17)8-10-19(18,20)2/h1,5,7,12,16-18,21-22H,4,6,8-11H2,2H3/t16-,17-,18+,19+,20-/m1/s1
Formel vikt 296.1776
Procent renhet >95%