Läs mer
17-Epiestriol, TRC
Produktkod. 30010334
Change view
Kvantitet:
2.5 mg
5 mg
10 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30010334 2.5 mg 2.5mg
30008963 10 mg -
30010978 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30010334 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE5865102.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1228-72-4
Molekylformel C18 H24 O3
Molekylvikt (g/mol) 288.38
Synonym Estra-1,3,5(10)-triene-3,16alpha,17alpha-triol, Estriol Imp. E (EP), 17-epi-Estriol, Estra-1,3,5(10)-triene-3,16,17-triol, (16α,17α)-, Estra-1,3,5(10)-triene-3,16α,17α-triol (7CI,8CI), (16α,17α)-Estra-1,3,5(10)-triene-3,16,17-triol, 16α,17α-Estriol, 16α-Hydroxy-17α-estradiol, 17-Epiestriol, 17α-Estriol, NSC 84051
IUPAC-namn (8R,9S,13S,14S,16R,17S)-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,16,17-triol
LEDER C[C@]12CC[C@H]3[C@@H](CCc4cc(O)ccc34)[C@@H]1C[C@@H](O)[C@H]2O
Specifikationer
Kemiskt namn eller material Estra-1,3,5(10)-triene-3,16alpha,17alpha-triol (17-epi-Estriol)
Anteckningar om renhetsgrad HPLC
Kvantitet 2.5 mg
InChI formel InChI=1S/C18H24O3/c1-18-7-6-13-12-5-3-11(19)8-10(12)2-4-14(13)15(18)9-16(20)17(18)21/h3,5,8,13-17,19-21H,2,4,6-7,9H2,1H3/t13-,14-,15+,16-,17-,18+/m1/s1
Formel vikt 288.1725
Procent renhet >95%