missing translation for 'onlineSavingsMsg'
Läs mer

17-Methyl-5Alpha-androst-1-ene-3Alpha,17Beta-diol, TRC

Produktkod. 30480729
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30480729 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30480729 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3272455MG

Please to purchase this item. Need a web account? Register with us today!

Denna vara kräver en prisuppdatering innan den beställs.

Begär ett pris för denna vara här.

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material 17-Methyl-5alpha-androst-1-ene-3alpha,17beta-diol
CAS 1173998-65-6
Anteckningar om renhetsgrad HPLC
Molekylformel C20 H32 O2
Kvantitet 5 mg
Synonym (3α,5α,17β)-17-Methyl-5-androst-1-ene-3,17-diol, 17α-Methyl-5α-androst-1-ene-3α,17β-diol
InChI formel InChI=1S/C20H32O2/c1-18-9-6-14(21)12-13(18)4-5-15-16(18)7-10-19(2)17(15)8-11-20(19,3)22/h6,9,13-17,21-22H,4-5,7-8,10-12H2,1-3H3/t13-,14+,15+,16-,17-,18-,19-,20-/m0/s1
LEDER C[C@]1(O)CC[C@H]2[C@@H]3CC[C@H]4C[C@H](O)C=C[C@]4(C)[C@H]3CC[C@]12C
IUPAC-namn (3S,5S,8R,9S,10R,13S,14S,17S)-10,13,17-trimethyl-3,4,5,6,7,8,9,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthrene-3,17-diol
Molekylvikt (g/mol) 304.47
Formel vikt 304.2402
Procent renhet >95
Visa mer Visa mindre
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.