Läs mer
17-Oxo Dexamethasone, TRC
Produktkod. 18083040
Change view
Kvantitet:
2.5 mg
5 mg
10 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
18083040 5 mg -
18010330 2.5 mg 2.5mg
18093630 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 18083040 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO8552005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1880-61-1
Molekylformel C20 H25 F O3
Molekylvikt (g/mol) 332.41
Synonym (11β,16α)-9-Fluoro-11-hydroxy-16-methylandrosta-1,4-diene-3,17-dione, Androsta-1,4-diene-3,17-dione, 9-fluoro-11β-hydroxy-16α-methyl- (7CI,8CI), (11β,16α)-9-Fluoro-11-hydroxy-16-methylandrosta-1,4-diene-3,17-dione, 9-Fluoro-11β-hydroxy-16α-methylandrosta-1,4-diene-3,17-dione, 9α-Fluoro-11β-hydroxy-16α-methylandrosta-1,4-diene-3,17-dione, NSC 50909
IUPAC-namn (8S,9R,10S,11S,13S,14S,16R)-9-fluoro-11-hydroxy-10,13,16-trimethyl-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthrene-3,17-dione
LEDER C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)C1=O
Specifikationer
Kemiskt namn eller material 17-Oxo Dexamethasone
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C20H25FO3/c1-11-8-15-14-5-4-12-9-13(22)6-7-19(12,3)20(14,21)16(23)10-18(15,2)17(11)24/h6-7,9,11,14-16,23H,4-5,8,10H2,1-3H3/t11-,14+,15+,16+,18+,19+,20+/m1/s1
Formel vikt 332.1788
Procent renhet >95%