Läs mer
17a-Methyl Testosterone, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | 17-alpha-Methyltestosterone |
| CAS | 58-18-4 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C20 H30 O2 |
| Kvantitet | 100 mg |
| Synonym | Methyltestosterone, 17alpha-Methyltestosterone, 17-Hydroxy-17-methyl-3-keto-androstene-4, 17-Methyltestosteron, 17-Methyltestosterone, Android, Andrometh, Androsan, Androsan (tablets), Androsten, Anertan, Anertan (tablets), CDB 110, Dumogran, Glosso-Sterandryl, Homandren, Hormale, L 589.372, M.T.Mucorettes, Malestrone, Malogen, Masenone, Mastestona, Mesterone, Metandren, Methyltestorone, Metrone, NSC 139965, NSC 9701, Neo-Hombreol-M, Nu-man, Oraviron, Orchisterone M, Oreton Methyl, Oreton-M, Perandren, RU 24400, Steronyl, Synandrets, Synandrotabs, Testhormone, Testora, Testoviron, Testoviron (tablet), Testovis Depot, Testred, U 2842, Androst-4-en-3-one, 17-hydroxy-17-methyl-, (17beta)-, Androst-4-en-3-one, 17beta-hydroxy-17-methyl- (8CI), (17beta)-17-Hydroxy-17-methylandrost-4-en-3-one, 17(alpha)-Methyl-Delta4-androsten-17-(beta)-ol-3-one, 17alpha-Methyl-17beta-hydroxyandrost-4-en-3-one, 17alpha-Methyl-17beta-hydroxyandrost-4-ene-3-one, 17alpha-Methyl-3-oxo-4-androsten-17beta-ol, 17alpha-Methylandrost-4-en-17-ol-3-one, 17alpha-Methylandrost-4-en-17beta-ol-3-one, 17beta-Hydroxy-17-methylandrost-4-en-3-one, 17beta-Hydroxy-17alpha-methyl-3-oxoandrost-4-ene, 17beta-Hydroxy-17alpha-methylandrost-4-en-3-one, 17beta-Hydroxy-17alpha-methylandrost-4-ene-3-one, Androst-4-ene-17alpha-methyl-17beta-ol-3-one |
| InChI formel | InChI=1S/C20H30O2/c1-18-9-6-14(21)12-13(18)4-5-15-16(18)7-10-19(2)17(15)8-11-20(19,3)22/h12,15-17,22H,4-11H2,1-3H3/t15-,16+,17+,18+,19+,20+/m1/s1 |
| LEDER | C[C@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C |
| IUPAC-namn | (8R,9S,10R,13S,14S,17S)-17-hydroxy-10,13,17-trimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-one |
| Molekylvikt (g/mol) | 302.45 |
| Visa mer |