Läs mer
17a-Oxa-D-homo-5beta-androst-1-ene-3,17-dione, TRC
Produktkod. 30009843
Change view
Kvantitet:
2.5 mg
5 mg
25 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30009843 5 mg -
30009475 25 mg -
30010837 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30009843 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO8707355MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4269-77-6
Molekylformel C19 H26 O3
Molekylvikt (g/mol) 302.41
Synonym (4aS,4bR,6aR,10aR,10bS,12aS)-3,4,4a,5,6,6a,7,10a,10b,11,12,12a-Dodecahydro-10a,12a-dimethyl-2H-phenanthro[2,1-b]pyran-2,8(4bH)-dione, 1-Phenanthrenepropionic acid, 1,2,3,4,4a,4b,7,8,8a,9,10,10a-dodecahydro-2-hydroxy-2,4b-dimethyl-7-oxo-, δ-lactone (7CI), 13,17-Seco-5β-androst-1-en-17-oic acid, 13-hydroxy-3-oxo-, δ-lactone (8CI), D-Homo-17a-oxaandrost-1-ene-3,17-dione, (5β)-, 1,2-Didehydro-4,5β-dihydrotestololactone, 17-Oxa-D-homo-5β-androst-1-en-3,17-dione, 17a-Oxa-D-homo-5β-androst-1-ene-3,17-dione
IUPAC-namn (4aS,4bR,6aR,10aR,10bS,12aS)-10a,12a-dimethyl-4,4a,4b,5,6,6a,7,10b,11,12-decahydro-3H-naphtho[2,1-f]chromene-2,8-dione
LEDER C[C@]12CC[C@H]3[C@@H](CC[C@@H]4CC(=O)C=C[C@]34C)[C@@H]1CCC(=O)O2
Specifikationer
Kemiskt namn eller material 17a-Oxa-D-homo-5beta-androst-1-ene-3,17-dione
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C19H26O3/c1-18-9-7-13(20)11-12(18)3-4-14-15(18)8-10-19(2)16(14)5-6-17(21)22-19/h7,9,12,14-16H,3-6,8,10-11H2,1-2H3/t12-,14-,15+,16+,18+,19+/m1/s1
Formel vikt 302.1882
Procent renhet >95%