Läs mer
(17Alpha)-13-Ethyl-3-methoxy-18,19-dinorpregna-3,5-dien-20-yn-17-ol, TRC
Produktkod. 30136781
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136781 100 mg
30005997 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136781 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE918945100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 176254-10-7
Molekylformel C22H30O2
Molekylvikt (g/mol) 326.47
Synonym 18,19-Dinorpregna-3,5-dien-20-yn-17-ol, 13-ethyl-3-methoxy-, (17α)-, 13-Ethyl-3-methoxy-18,19-dinor-17α-pregna-3,5-dien-20-yn-17-ol, Levonorgestrel Imp. S
IUPAC-namn (8R,9S,10R,13S,14S,17R)-13-ethyl-17-ethynyl-3-methoxy-2,7,8,9,10,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-ol
LEDER CC[C@]12CC[C@H]3[C@@H](CC=C4C=C(CC[C@H]34)OC)[C@@H]1CC[C@@]2(O)C#C
Specifikationer
Kemiskt namn eller material (17alpha)-13-Ethyl-3-methoxy-18,19-dinorpregna-3,5-dien-20-yn-17-ol
Kvantitet 100 mg
InChI formel InChI=1S/C22H30O2/c1-4-21-12-10-18-17-9-7-16(24-3)14-15(17)6-8-19(18)20(21)11-13-22(21,23)5-2/h2,6,14,17-20,23H,4,7-13H2,1,3H3/t17-,18+,19+,20-,21-,22-/m0/s1
Formel vikt 326.2246