Läs mer
(17alpha)-19-Norpregna-3,5-dien-20-yne-3,17-diol 3,17-Diacetate, TRC
Produktkod. 30416738
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30416738 100 mg
1 options
Produktkod. 30416738 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN825020100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (17alpha)-19-Norpregna-3,5-dien-20-yne-3,17-diol 3,17-Diacetate
CAS 2205-78-9
Molekylformel C24 H30 O4
Kvantitet 100 mg
Synonym (17α)-19-Norpregna-3,5-dien-20-yne-3,17-diol Diacetate, 17-Ethynyl-19-norandrosta-3,5-diene-3,17β-diol Diacetate, 19-Nor-17α-pregna-3,5-dien-20-yne-3,17β-diol Diacetate, SC 6091
InChI formel InChI=1S/C24H30O4/c1-5-24(28-16(3)26)13-11-22-21-8-6-17-14-18(27-15(2)25)7-9-19(17)20(21)10-12-23(22,24)4/h1,6,14,19-22H,7-13H2,2-4H3/t19-,20+,21+,22-,23-,24-/m0/s1
LEDER CC(=O)OC1=CC2=CC[C@@H]3[C@H](CC[C@@]4(C)[C@H]3CC[C@@]4(OC(=O)C)C#C)[C@H]2CC1
IUPAC-namn [(8R,9S,10R,13S,14S,17R)-17-acetyloxy-17-ethynyl-13-methyl-2,7,8,9,10,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-3-yl] acetate
Molekylvikt (g/mol) 382.49
Formel vikt 382.2144