Learn More
17Alpha-Boldenone 17-O-(4-Nitrobenzoate), TRC
Specifications
Specifications
| Chemical Name or Material | 17alpha-Boldenone 17-O-(4-Nitrobenzoate) |
| Analyte or Component Names | 17alpha-Boldenone 17-O-(4-Nitrobenzoate) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C26H29NO5 |
| Synonym | (17α)-17-Hydroxyandrosta-1,4-dien-3-one 17-O-(4-Nitrobenzoate) |
| Solution Type | Neat |
| SMILES | O=C1C=C[C@@]2(C)C(CC[C@@]([H])([C@@]34[H])[C@]2([H])CC[C@@]3([C@H](OC(C5=CC=C([N+]([O-])=O)C=C5)=O)CC4)C)=C1 |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 435.51 |
| Quantity | 5 mg |