Läs mer
17Alpha-Hydroxyprogesterone Valerate, TRC
Produktkod. 30133939
Change view
Kvantitet:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133939 500 mg
30005234 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133939 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH961915500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6677-15-2
Molekylformel C26 H38 O4
Molekylvikt (g/mol) 414.58
Synonym 17α-Hydroxyprogesterone Valerate, 17-Hydroxypregn-4-ene-3,20-dione Valerate
IUPAC-namn [(8R,9S,10R,13S,14S,17R)-17-acetyl-10,13-dimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl] pentanoate
LEDER CCCCC(=O)O[C@@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C)C(=O)C
Specifikationer
Kemiskt namn eller material 17 alpha-Hydroxyprogesterone Valerate
Kvantitet 500 mg
InChI formel InChI=1S/C26H38O4/c1-5-6-7-23(29)30-26(17(2)27)15-12-22-20-9-8-18-16-19(28)10-13-24(18,3)21(20)11-14-25(22,26)4/h16,20-22H,5-15H2,1-4H3/t20-,21+,22+,24+,25+,26+/m1/s1
Formel vikt 414.277