missing translation for 'onlineSavingsMsg'
Läs mer

17Alpha-Trenbolone-d3, TRC

Product Code. 30452717
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Kvantitet:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30452717 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30452717 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCT71899310MG

Please to purchase this item. Need a web account? Register with us today!

Denna vara kräver en prisuppdatering innan den beställs.

Begär ett pris för denna vara här.

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 17alpha-Trenbolone 16,16,17-d3
CAS 2026643-10-5
Anteckningar om renhetsgrad HPLC
Molekylformel C18 2H3 H19 O2
Kvantitet 10 mg
Synonym (17α)-17-Hydroxyestra-4,9,11-trien-3-one-16,16,17-d3, 17-Epitrenbolone-16,16,17-d3, 17α-Trenbolone-16,16,17-D3, Epitrenbolone-d3, α-Trenbolone-d3, 17α-Trenbolone-d3
InChI formel InChI=1S/C18H22O2/c1-18-9-8-14-13-5-3-12(19)10-11(13)2-4-15(14)16(18)6-7-17(18)20/h8-10,15-17,20H,2-7H2,1H3/t15-,16+,17-,18+/m1/s1/i7D2,17D
LEDER [2H]C1([2H])C[C@H]2[C@@H]3CCC4=CC(=O)CCC4=C3C=C[C@]2(C)[C@]1([2H])O
IUPAC-namn (8S,13S,14S,17R)-16,16,17-trideuterio-17-hydroxy-13-methyl-2,6,7,8,14,15-hexahydro-1H-cyclopenta[a]phenanthren-3-one
Molekylvikt (g/mol) 273.38
Formel vikt 273.1808
Procent renhet >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.