Läs mer
17Beta-Acetyl-estradiol 3-(Tri-O-acetyl-Beta-D-glucuronic Acid Methyl Ester), TRC
Produktkod. 30321959
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30321959 10 mg
1 options
Produktkod. 30321959 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA17382510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 17beta-Acetyl-estradiol 3-(Tri-O-acetyl-beta-D-glucuronic Acid Methyl Ester)
CAS 59495-70-4
Molekylformel C33H42O12
Kvantitet 10 mg
Synonym (17beta)-17-Acetoxyestra-1,3,5(10)-trien-3-yl Tri-O-acetyl-beta-D-glucuronic Acid Methyl Ester, 17-Acetyl-estradiol Tri-O-acetyl-beta-D-glucuronic Acid Methyl Ester
InChI formel InChI=1S/C33H42O12/c1-16(34)40-26-12-11-25-24-9-7-20-15-21(8-10-22(20)23(24)13-14-33(25,26)5)44-32-30(43-19(4)37)28(42-18(3)36)27(41-17(2)35)29(45-32)31(38)39-6/h8,10,15,23-30,32H,7,9,11-14H2,1-6H3/t23-,24-,25+,26+,27+,28+,29+,30-,32-,33+/m1/s1
LEDER COC(=O)[C@H]1O[C@@H](Oc2ccc3[C@H]4CC[C@]5(C)[C@H](CC[C@H]5[C@@H]4CCc3c2)OC(=O)C)[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@@H]1OC(=O)C
IUPAC-namn methyl (2S,3S,4S,5R,6S)-3,4,5-triacetyloxy-6-[[(8R,9S,13S,14S,17S)-17-acetyloxy-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-3-yl]oxy]oxane-2-carboxylate
Molekylvikt (g/mol) 630.68
Formel vikt 630.2676