missing translation for 'onlineSavingsMsg'
Läs mer

17Beta-Acetyloxy-5Alpha-androstenone, TRC

Produktkod. 30328662
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30328662 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30328662 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCA16433025MG

Please to purchase this item. Need a web account? Register with us today!

Denna vara kräver en prisuppdatering innan den beställs.

Begär ett pris för denna vara här.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material 17beta-Acetyloxy-5alpha-androstenone
CAS 64-82-4
Molekylformel C21H30O3
Kvantitet 25 mg
Synonym (5alpha,17beta)-17-(Acetyloxy)-androst-1-en-3-one, 17beta-Acetoxy-5alpha-androst-1-en-3-one, 17beta-Hydroxy-5alpha-androst-1-en-3-one acetate, ZK 5777
InChI formel InChI=1S/C21H30O3/c1-13(22)24-19-7-6-17-16-5-4-14-12-15(23)8-10-20(14,2)18(16)9-11-21(17,19)3/h8,10,14,16-19H,4-7,9,11-12H2,1-3H3/t14-,16-,17-,18-,19-,20-,21-/m0/s1
LEDER CC(=O)O[C@H]1CC[C@H]2[C@@H]3CC[C@H]4CC(=O)C=C[C@]4(C)[C@H]3CC[C@]12C
IUPAC-namn [(5S,8R,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-4,5,6,7,8,9,11,12,14,15,16,17-dodecahydrocyclopenta[a]phenanthren-17-yl] acetate
Molekylvikt (g/mol) 330.46
Formel vikt 330.2195
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.