Läs mer
17Beta-Carboxy-17-desoxy Dexamethasone, TRC
Produktkod. 17994255
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17994255 25 mg
17993535 10 mg
17944795 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17994255 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC17792525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 75262-69-0
Molekylformel C21 H27 F O4
Molekylvikt (g/mol) 362.44
Synonym Androsta-1,4-diene-17-carboxylic acid, 9-fluoro-11-hydroxy-16-methyl-3-oxo-, (11β,16α,17β)-, (11β,16α,17β)-9-Fluoro-11-hydroxy-16-methyl-3-oxoandrosta-1,4-diene-17-carboxylic acid, Dexamethasone-17α-dehydroxy-17β-carboxylic acid, 9-Fluoro-11β-hydroxy-16α-methyl-3-oxoandrosta-1,4-diene-17β-carboxylic acid, Desoximetasone acid, 17β-Carboxy-17-desoxy Dexamethasone
IUPAC-namn (8S,9R,10S,11S,13S,14S,16R,17S)-9-fluoro-11-hydroxy-10,13,16-trimethyl-3-oxo-7,8,11,12,14,15,16,17-octahydro-6H-cyclopenta[a]phenanthrene-17-carboxylic acid
LEDER C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@H]1C(=O)O
Specifikationer
Kemiskt namn eller material 17beta-Carboxy-17-desoxy Dexamethasone
Kvantitet 25 mg
InChI formel InChI=1S/C21H27FO4/c1-11-8-15-14-5-4-12-9-13(23)6-7-20(12,3)21(14,22)16(24)10-19(15,2)17(11)18(25)26/h6-7,9,11,14-17,24H,4-5,8,10H2,1-3H3,(H,25,26)/t11-,14+,15+,16+,17-,19+,20+,21+/m1/s1
Formel vikt 362.1893