Läs mer
17Beta-Carboxy-17Alpha-formyloxy Dexamethasone, TRC
Produktkod. 30004698
Change view
Kvantitet:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30004698 5 mg
30003634 25 mg
17923415 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30004698 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC1784305MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 473273-04-0
Molekylformel C22 H27 F O6
Molekylvikt (g/mol) 406.44
Synonym Androsta-1,4-diene-17-carboxylic acid, 9-fluoro-17-(formyloxy)-11-hydroxy-16-methyl-3-oxo-, (11β,16α,17α)- (9CI), 17β-Carboxy-17α-formyloxydexamethasone
IUPAC-namn (8S,9R,10S,11S,13S,14S,16R,17R)-9-fluoro-17-formyloxy-11-hydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthrene-17-carboxylic acid
LEDER C[C@@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(OC=O)C(=O)O
Specifikationer
Kemiskt namn eller material 17beta-Carboxy-17alpha-formyloxy Dexamethasone
Anteckningar om renhetsgrad HPLC
Kvantitet 5 mg
InChI formel InChI=1S/C22H27FO6/c1-12-8-16-15-5-4-13-9-14(25)6-7-19(13,2)21(15,23)17(26)10-20(16,3)22(12,18(27)28)29-11-24/h6-7,9,11-12,15-17,26H,4-5,8,10H2,1-3H3,(H,27,28)/t12-,15+,16+,17+,19+,20+,21+,22+/m1/s1
Formel vikt 406.1792
Procent renhet >95%