Läs mer
17beta-Dihydro Equilenin 3-Sulfate Sodium Salt (Stabilized with TRIS, 50% w/w), TRC
Produktkod. 30376017
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30376017 25 mg
1 options
Produktkod. 30376017 Leverantör Toronto Research Chemicals Leverantörsnummer TRCD44896525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 17Beta-Dihydroequilenin Sodium Sulfate
CAS 16680-50-5
Molekylformel C18 H19 O5 S . Na
Kvantitet 25 mg
Synonym Estra-1,3,5,7,9-pentaene-3,17-diol, 3-(hydrogen sulfate), sodium salt (1:1), (17β)-, Estra-1,3,5,7,9-pentaene-3,17-diol, 3-(hydrogen sulfate), monosodium salt, (17β)- (9CI), Estra-1,3,5,7,9-pentaene-3,17β-diol, 3-(hydrogen sulfate), sodium salt (8CI), 17β
InChI formel InChI=1S/C18H20O5S.Na/c1-18-9-8-14-13-5-3-12(23-24(20,21)22)10-11(13)2-4-15(14)16(18)6-7-17(18)19;/h2-5,10,16-17,19H,6-9H2,1H3,(H,20,21,22);/q;+1/p-1/t16-,17-,18-;/m0./s1
LEDER [Na+].C[C@]12CCc3c(ccc4cc(OS(=O)(=O)[O-])ccc34)[C@@H]1CC[C@@H]2O
IUPAC-namn sodium;[(13S,14S,17S)-17-hydroxy-13-methyl-11,12,14,15,16,17-hexahydrocyclopenta[a]phenanthren-3-yl] sulfate
Molekylvikt (g/mol) 370.395
Formel vikt 370.085