Läs mer
(17Beta)-Estra-1,3,5(10),6,8-pentaene-3,17Beta-diol Diacetate, TRC
Produktkod. 30372745
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30372745 25 mg
1 options
Produktkod. 30372745 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE66863525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (17beta)-Estra-1,3,5(10),6,8-pentaene-3,17beta-diol Diacetate
CAS 1423-96-7
Molekylformel C22H24O4
Kvantitet 25 mg
Synonym (13S,14S,17S)-13-methyl-12,13,14,15,16,17-Hexahydro-11H-cyclopenta[a]phenanthrene-3,17-diyl Diacetate, Estra-1,3,5(10),6,8-pentaene-3,17β-diol Diacetate
InChI formel InChI=1S/C22H24O4/c1-13(23)25-16-5-7-17-15(12-16)4-6-19-18(17)10-11-22(3)20(19)8-9-21(22)26-14(2)24/h4-7,12,20-21H,8-11H2,1-3H3/t20-,21-,22-/m0/s1
LEDER C[C@]12CCC3=C([C@]1([H])CC[C@@H]2OC(C)=O)C=CC4=C3C=CC(OC(C)=O)=C4
IUPAC-namn (13S,14S,17S)-13-methyl-12,13,14,15,16,17-hexahydro-11H-cyclopenta[a]phenanthrene-3,17-diyl diacetate
Molekylvikt (g/mol) 352.42
Formel vikt 352.17