Läs mer
17Beta-Estradiol 3-Beta-D-Glucuronide, TRC
Produktkod. 30011479
Change view
Kvantitet:
2.5 mg
25 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30011479 25 mg -
30005042 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30011479 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE88801025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 15270-30-1
Molekylformel C24H32O8
Molekylvikt (g/mol) 448.51
Synonym (17β)-17-Hydroxyestra-1,3,5(10)-trien-3-yl β-D-Glucopyranosiduronic Acid, 3-(β-D-Glucopyranuronosyloxy) Estra-1,3,5(10)-trien-17β-ol, Estradiol 3-Glucuronide;
LEDER C[C@]12CC[C@H]3[C@H]([C@@H]1CC[C@@H]2O)CCC4=C3C=CC(=C4)O[C@H]5[C@@H]([C@H]([C@@H]([C@H](O5)C(=O)O)O)O)O
Specifikationer
Kemiskt namn eller material 17beta-Estradiol 3-beta-D-Glucuronide
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C24H32O8/c1-24-9-8-14-13-5-3-12(10-11(13)2-4-15(14)16(24)6-7-17(24)25)31-23-20(28)18(26)19(27)21(32-23)22(29)30/h3,5,10,14-21,23,25-28H,2,4,6-9H2,1H3,(H,29,30)/t14-,15-,16+,17+,18+,19+,20-,21+,23-,24+/m1/s1
Procent renhet >95%