Läs mer
17beta-Hydroxyestra-1,3,5(10)-trien-3-yl Pentanoate (Estradiol 3-Valerate), Mikromol™
Produktkod. 30049219
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30049219 25 mg
30049737 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30049219 Leverantör Mikromol Leverantörsnummer MM0209.060025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
17beta-Hydroxyestra-1,3,5(10)-trien-3-yl Pentanoate (Estradiol 3-Valerate)
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Estradiol 3-Valerate
CAS 21881-45-8
Analyt- eller komponentnamn Estradiol 3-Valerate
Frakt skick Room Temperature
Molekylformel C23 H32 O3
Certifikat och efterlevnad ISO 17025
Synonym 17beta-Hydroxyestra-1,3,5(10)-trien-3-yl Pentanoate, Estradiol Valerate Imp. B (EP), Estradiol 3-Valerate
InChI formel InChI=1S/C23H32O3/c1-3-4-5-22(25)26-16-7-9-17-15(14-16)6-8-19-18(17)12-13-23(2)20(19)10-11-21(23)24/h7,9,14,18-21,24H,3-6,8,10-13H2,1-2H3/t18-,19-,20+,21+,23+/m1/s1
LEDER CCCCC(=O)Oc1ccc2[C@H]3CC[C@]4(C)[C@@H](O)CC[C@H]4[C@@H]3CCc2c1
IUPAC-namn [(8R,9S,13S,14S,17S)-17-hydroxy-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-3-yl] pentanoate
Molekylvikt (g/mol) 356.51
Kvantitet 25 mg
Formel vikt 356.506 g/mol