Learn More
Bochem™ 18/10 E-Poli Silicone Beaker Tongs
Product Code. 10741621
Change view
Click to view available options
Length (Metric) Opening:
140 mm
150 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Opening Unit Size
10741621 150 mm Each
10300891 140 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10741621 Supplier Bochem™ Supplier No. 2200
Top Sellers. Mega Savings.
Use promo code 27997
View all promotions
Beräknad leverans: 01-02-2027
Specifications
Product Type Beaker Tongs
Length (Metric) Opening 150 mm
Length (Metric) 250 mm
Material Stainless Steel (18/10)